Baculovirus expression system, flashBAC™
Lieferant: MIRUS
MIR6115
MIR6120
MIR6135
MIR6140
MIRUMIR6115EA
513
EUR
MIRUMIR6115
MIRUMIR6120
MIRUMIR6135
MIRUMIR6140
Baculovirus expression system, flashBAC™
Transfektionsreagenzien
The flashBAC™ Baculovirus Expression System is designed for high yield protein production in insect and mammalian cells.
- Single-step baculovirus generation in insect cells, no plaque purification required
- Ready to use with most transfer vectors
- High recombinant protein yields
This system is a streamlined platform for the production of recombinant baculoviruses. Genetic optimisation of the Autographa californica nucleopolyhedrovirus (AcMNPV) genome yields recombinant virus in a single step and eliminates the need for separation of recombinant and parental virus by plaque purification.