In vivo expression and reporter vectors, pLIVE®
Lieferant: MIRUS
MIR5320 MIR5420 MIR5520 MIR5620
MIRUMIR5320EA 1230 EUR
MIRUMIR5320 MIRUMIR5420 MIRUMIR5520 MIRUMIR5620
In vivo expression and reporter vectors, pLIVE®
Transfektionsreagenzien
pLIVE®
  • Achieve sustained gene expression – long term liver gene expression in mouse liver through 8 months post-injection
  • Available with or without positive control vectors expressing either lacZ or SEAP