Adenosin-5'-monophosphorsäure (AMP) Monohydrat freie Säure (aus Hefe), weißes Pulver
Lieferant: MP Biomedicals
Synonyme: A-5′-P, AMP, Adenosine-5'-monophosphoric acid monohydrate, 5′-AMP, 5′-Adenylic acid
0210008080 0210008001 0210008025
ICNA0210008080EA 558 EUR
ICNA0210008080 ICNA0210008001 ICNA0210008025
Adenosin-5'-monophosphorsäure (AMP) Monohydrat freie Säure (aus Hefe), weißes Pulver
Adenosin-5'-monophosphorsäure (AMP) Monohydrat
Adenosine 5′-monophosphate is a nucleotide that is used as a monomer in RNA. It is an ester of phosphoric acid and the nucleoside adenosine. AMP consists of a phosphate group, the sugar ribose, and the nucleobase adenine.
Adenosine 5′-monophosphate (5′-AMP) has many uses in nature. 5′-AMP is an activator of a class of protein kinases known as AMP-activated protein kinase (AMPK).
It has many uses in nature, 5′-AMP is an activator of a class of protein kinases known as AMP-activated protein kinase (AMPK).
Soluble in water with addition of mild alkali, 1N ammonium hydroxide (200 mg/4 ml-clear to slightly hazy colorless solution).
Bitte beachten Research Use Only (RUO).
Formel: C₁₀H₁₄N₅O₇P·1H₂O
Molecular Weight: 365.24 g/mol
MDL: MFCD00149360
CAS-Nummer: 18422-05-4
Bestellen
Spezifikation Testergebnisse
Identity Test Passes
Source Yeast
FTIR Conforms to Standard
UV/Vis: λmax (water) 258 ± 5 n
OD250/OD260 0.83 ± 0.0
OD 280/OD26 0.15 ± 0.0
Loss on Drying <7%