Spermin Tetrahydrochlorid ≥96%, weißes Pulver
Lieferant: MP Biomedicals
|
Achtung
|
Synonyme:
N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine) tetrahydrochloride
Spermine is a naturally occuring polyamine that occurs in all eukaryotes, but is rare in prokaryotes. It is essential for cell growth in both normal and neoplastic tissue.
Spermine is formed through the addition of an aminopropyl group to spermidine by spermine synthase. Spermine is strongly basic in character, and in aqueous solution at physiological pH, all of its amino groups will be positively charged.
Used to precipitate DNA from low salt aqueous buffers. Spermine tetrahydrochloride was used to precipitate DNA for flow cytometric analysis from fresh frozen breast cancer tissue, archival tissue of salivary gland, renal and thyroid tumors and red blood cells of chicken.
Mixed NMDA agonist/antagonist at the polyamine site. Neuroprotective effects have been observed at high concentrations (1 mM), while neurotoxicity is observed at lower concentrations. It enhances agonist effectiveness at the strychnine-insensitive glycine site.
Soluble in water (100 mg/mL - clear, colorless solution).
Bitte beachten Research Use Only (RUO).
Formel:
C₁₀H₃₀Cl₄N₄ Molecular Weight: 348.19 g/mol Schmelzpunkt: 312…314 °C (Lit.) Lagertemperatur: Gefrierschrank |
MDL:
MFCD00012914 CAS-Nummer: 306-67-2 |
Spezifikation Testergebnisse
Identity Test | Passes |
Purity | ≥96% |
FTIR | Conforms to Standard |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...