(±)-Asparaginsäure ≥99%, weißes Pulver
Lieferant: MP Biomedicals
|
Achtung
|
Synonyme:
2-Aminobutanedioic acid, DL-Aspartic acid
0210080383
0210080390
0210080391
ICNA0210080383EA
43.9
EUR
ICNA0210080383
ICNA0210080390
ICNA0210080391
(±)-Asparaginsäure ≥99%, weißes Pulver
(±)-Asparaginsäure
DL-Aspartic acid (DL-Asp) is a racemic mixture of the proteinogenic amino acid L-aspartate and the non-proteinogenic amino acid D-aspartate. DL-Aspartic acid is used in studies on factors and conditions that enhance semen and sperm quality.
L-Aspartic acid is an endogenous NMDA receptor agonist. Principal neurotransmitter for fast synaptic excitation.
Soluble in NaOH 100 mg/ml.
Bitte beachten Research Use Only (RUO).
Formel:
C₄H₇NO₄ Molecular Weight: 133,10 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063083 CAS-Nummer: 617-45-8 |
Spezifikation Testergebnisse
Identity Test | Passes |
Purity | ≥99% |
FTIR | Conforms to Standard |
Optical Rotation (c=1, 1N HCl) |
+21 to +27° |
Loss on Drying | ≤0.5% |
pH | 3,0 (Lit.) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...