L(-)-Cystin ~99%, kristallin
Lieferant: MP Biomedicals
Synonyme:
(R)-(-)-Cystin, (2R,2'R)-(-)-3,3'-Dithiobis(2-aminopropansäure), (H-Cys-OH)2
0210145425
0210145480
0210145490
0210145491
ICNA0210145425EA
36.1
EUR
ICNA0210145425
ICNA0210145480
ICNA0210145490
ICNA0210145491
L(-)-Cystin ~99%, kristallin
L(-)-Cystin
50 mg/ml, 1 M HCl.
Solubility: 50 mg/ml, 1 M HCl.
Bitte beachten Research Use Only (RUO).
Formel:
HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH Molecular Weight: 240,3 g/mol Schmelzpunkt: >240 °C (Zers., Lit.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064228 CAS-Nummer: 56-89-3 |
Spezifikation Testergebnisse
Identity Test | Passes |
Purity | 98.5 - 101.0% |
FTIR | Conforms to Standard |
Moisture | ≤0.5% |
Optical Rotation (c=1,1N HCl) | –210 to –230° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...