Hämatoxylin Hydrat, orangenes Pulver C.I. 75290

Lieferant: MP Biomedicals
Achtung

Synonyme: (6aS,11bR)-7,11b-Dihydro-6H-indeno[2,1-c]chromen-3,4,6a,9,10-pentol Hydrat, Hydroxybrasilin Hydrat, Naturschwarz 1, (+)-Hämatoxylin Hydrat

0210192205 0210192225 0210192280
ICNA0210192205EA 44.8 EUR
ICNA0210192205 ICNA0210192225 ICNA0210192280
Hämatoxylin Hydrat, orangenes Pulver C.I. 75290
Hämatoxylin Hydrat

Hematoxylin is Gill’s Formulation 2; a nuclear counter stain of blue-violet colour. It is extracted from the heartwood of the logwood tree.


When oxidized it forms haematein, a compound that forms strongly coloured complexes with certain metal ions, the most notable ones being Fe(III) and Al(III) salts. Metal-haematein complexes are used to stain cell nuclei prior to examination under a microscope. Structures that stain with iron- or aluminium-haematein are often called basophilic, even though the mechanism of the staining is different from that of staining with basic dyes.


Used as a nuclear stain in cytology and histology applications.


Soluble in ethanol (20 mg/ml); hot water.

Formel: C₁₆H₁₄O₆·xH₂O
Molecular Weight: 302.29 g/mol (anhy)
Schmelzpunkt: 229…231 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00078111
CAS-Nummer: 517-28-2

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
UV/VIS: l max (meOH) 290 - 292 nm
Loss on Drying Report Result


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR