Eusolex® 9020
Lieferant: Merck
|
|
1.05844.9027
1.05844.0101
1.05844.1001
1.05844.9027EA
0
EUR
1.05844.9027
1.05844.0101
1.05844.1001
Eusolex® 9020
Eusolex®
Organic UV Filter
Eusolex® 9020 is the most effective UV-A filter, globally approved and of wide-spread use in many formulations. Eusolex® 9020 is an off-white powder and needs to be solublized and photostabilized in the oil phase.
INCI name: Butyl Methoxydibenzoylmethane
Formel:
C₂₀H₂₂O₃ Schmelzpunkt: 81…86 °C Dichte: 1,22 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
CAS-Nummer:
70356-09-1 EINECS: 274-581-6 |
Spezifikation Testergebnisse
Synonyms | 1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl) 1,3-propanedione) |
Storage | Storage temperature: no restrictions |
Solubility | 0,027 g/l |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...