4-(3-Indolyl)butansäure, weißgraues Puder

Lieferant: MP Biomedicals
Gefahr

Synonyme: 4-Indol-3-buttersäure, 4-(Indol-3-yl)buttersäure

0210204201 0210204205 0210204225 0210204250
ICNA0210204201EA 33.4 EUR
ICNA0210204201 ICNA0210204205 ICNA0210204225 ICNA0210204250
4-(3-Indolyl)butansäure, weißgraues Puder
4-(3-Indolyl)butansäure

Indole-3-butyric acid is used as plant growth hormone.


A naturally occurring phytohormone auxin (plant growth regulator). Promotes root formation in cuttings, but does not affect ethylene levels.


Bitte beachten Research Use Only (RUO).

Formel: C₁₂H₁₃NO₂
Molecular Weight: 203,24 g/mol
MDL: MFCD00005664
CAS-Nummer: 133-32-4
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard
Melting Point 118 - 126 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR