β-Sitosterol, powder_white, Practical

Lieferant: MP Biomedicals

Synonyme: α-Dihydrofucosterol, β-Sitosterol, 5-Stigmasten-3β-ol, 22,23-Dihydrostigmasterol, 24α-Ethylcholesterol

0210288610 0210288680 0210288690
ICNA0210288610EA 37.7 EUR
ICNA0210288610 ICNA0210288680 ICNA0210288690
β-Sitosterol, powder_white, Practical
beta-Sitosterol

β-Sitosterol has recently been shown to induce G2/M arrest, endoreduplication, and apoptosis through the Bcl-2 and PI3K/Akt signaling pathways.


Soluble in chloroform (50 mg/ml - clear, colorless solution).


Bitte beachten Research Use Only (RUO).

Formel: C₂₉H₅₀O
Molecular Weight: 414.72 g/mol
MDL: MFCD00003631
CAS-Nummer: 83-46-5

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Source Soybeans
beta-Sitosterol 40 - 60%
Campesterol 20 - 40%
Stigmasterol ≥5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR