L-Pipecolsäure ≥99%, weißes kristallines Pulver

Lieferant: MP Biomedicals

Synonyme: (S)-(-)-2-Piperidinecarboxylic Acid, H-HomoPro-OH, (S)-Piperidine-2-carboxylic acid, L-Pipecolinic Acid, L-Homoproline

0215189825 0215189880 0215189883 0215189891
ICNA0215189825EA 36.7 EUR
ICNA0215189825 ICNA0215189880 ICNA0215189883 ICNA0215189891
L-Pipecolsäure ≥99%, weißes kristallines Pulver
L-Pipecolsäure

L-pipecolic acid (L-PA) is use as an important biochemical marker for the diagnosis of peroxisomal disorders.


L-Pipecolic acid is a proline homolog and lysine metabolite. It occurs in seeds, malt, edible mushrooms, fruits, etc. Defects in its metabolism is involved in hyperpipecolic acidemia, cerebro-hepato-renal syndrome, neonatal onset adrenoleukodystrophy, and infantile Refsum disease.


Soluble in 50% methanol (50 mg/mL-clear, colorless solution) or water (50 mg/mL - clear to slightly hazy, colorless to faint yellow solution); sparingly soluble in acetone, chloroform; insoluble in ether.

Formel: C₆H₁₁NO₂
Molecular Weight: 129,16 g/mol
MDL: MFCD00005981
CAS-Nummer: 3105-95-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR