Nε-Acetyl-L-lysin ≥99%, Pulver

Lieferant: MP Biomedicals

Synonyme: Nε-Lys(Ac)-OH, (S)-6-Acetamido-2-aminohexanoic acid, L-Lys(Ac)-OH, H-Lys(Ac)-OH, Nepsilon-acetyl-L-lysine

02150235.2 0215023501 0215023505
ICNA02150235.2EA 39.8 EUR
ICNA02150235.2 ICNA0215023501 ICNA0215023505
Nε-Acetyl-L-lysin ≥99%, Pulver
Nε-Acetyl-L-lysin

Soluble in 80% AcOH (5 mg/ml - clear, colorless solution) and water.


Standards: Soluble in 80% AcOH (5 mg/ml - clear, colorless solution) and water.


Bitte beachten Research Use Only (RUO).

Formel: C₈H₁₆N₂O₃
Molecular Weight: 188,23 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00002639
CAS-Nummer: 692-04-6

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
pH (10% solution) 7.0 - 9.7

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR