D(+)-Mannose ≥98%, weißes kristallines Pulver

Lieferant: MP Biomedicals

Synonyme: (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal

0210225025 0210225080
ICNA0210225025EA 44.9 EUR
ICNA0210225025 ICNA0210225080
D(+)-Mannose ≥98%, weißes kristallines Pulver
D(+)-Mannose

Mannose is a sugar monomer of the aldohexose series of carbohydrates.


D-(+)-Mannose, a C-2 epimer of D-glucose, is used in the formation of glycan structure and glycosylation. D-Mannose has been used in a study to assess the synthesis of a family of amphiphilic glycopolymers. It has also been used in a study to investigate the early detection of bronchiolitis obliterans after lung transplantation.


D-mannose is sold as a naturopathic remedy for urinary tract infections, and it is claimed to work through the disruption of adherence of bacteria in the urinary tract. It can also be used to study the effects of exogenous mannose on in vitro processes such as glycosylation.


Soluble in water (50 g/ml - clear colorless solution).


Bitte beachten Research Use Only (RUO).

Formel: C₆H₁₂O₆
Molecular Weight: 180.16 g/mol
Siedepunkt: 376
Schmelzpunkt: 133
Lagertemperatur: Raumtemperatur
MDL: MFCD00064122
CAS-Nummer: 3458-28-4
Merck Index: 13,05772

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥98%
FTIR Conforms to standard
Melting Point 129 - 140 °C
Loss on Drying ≤0.5%
Optical Rotation (c=4, H₂O, 24 hour) +14.5±1°
Heavy Metals ≤10 ppm
Residue on Ignition ≤0.1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR