3,5-Dihydroxytoluol Monohydrat ≥98%, hellbeiges Pulver
Lieferant: MP Biomedicals
|
Achtung
|
Synonyme:
Orcin Monohydrat, 5-Methylresorcin Monohydrat
0210250650
0210250610
ICNA0210250650EA
217
EUR
ICNA0210250650
ICNA0210250610
3,5-Dihydroxytoluol Monohydrat ≥98%, hellbeiges Pulver
3,5-Dihydroxytoluol Monohydrat
Orcinol is used to prepare colorimetric reagents for the determination of RNA, DNA, hexoses and pentoses.
Orcinol is used to prepare colorimetric reagents for the determination of RNA, DNA, hexoses and pentoses.
Solubility : Soluble in alcohol and water (5 mg/ mL-clear colorless to faint yellow solution).
Bitte beachten Storage: protect from light. Research Use Only (RUO).
Formel:
C₇H₈O₂·1H₂O Molecular Weight: 142,15 g/mol Schmelzpunkt: 57,..67 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149092 CAS-Nummer: 6153-39-5 |
Spezifikation Testergebnisse
Identity Test | Passes |
Purity | >98% |
FTIR | Conforms to Standard |
Melting Point | 57 - 67°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...