Rubidiumchlorid ≥99%, weißes Pulver, Optische Qualität
Lieferant: MP Biomedicals
Synonyme:
Rubidiumchloride
Rubidium chloride is used as a catalyst and additive in gasoline.
In materials science, RbCl has been used to prepare molecular nanowires as potential precursors of nanoscale devices. In chromatography, RbCl and other univalent salts have been studied for their influence on the capillary electrophoretic separation of amino acids labeled with 3-(4-carboxybenzoyl)-quinoline-2-carboxaldehyde. RbCl is utilised in the transfection of bacteria. RbCl has been used to investigate the gating and permeability of ion channels produced by botulinum neurotoxin types A and E in membranes from cultured PC12 cells.
Formel:
RbCl Molecular Weight: 120,92 g/mol Schmelzpunkt: 715 °C (Lit.) |
MDL:
MFCD00011187 CAS-Nummer: 7791-11-9 |
Spezifikation Testergebnisse
Identity Test | Passes |
Purity | ≥99% |
OD280 (40% aq soln) | ≤0,2 |
Solubility (40% aq soln) | Clear, Colorless Solution |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...