O15-Palmitoylretinol, hellgelbes Pulver

Lieferant: MP Biomedicals

Synonyme: Retinyl palmitate, Retinol palmitate, alle-trans-Retinylpalmitat

0210326210 0210326280
ICNA0210326210EA 65.5 EUR
ICNA0210326210 ICNA0210326280
O15-Palmitoylretinol, hellgelbes Pulver
Vitamin A palmitate

Vitamin A palmitate, type P 1,7 is a pure, high potency form of all- trans retinyl palmitate. This product is prepared without antioxidants.


This product is the dry form of Vitamin A palmitate and is dispersable in water. However, the pure Viamin A palmitate is miscible with edible oils and fats; soluble in both hydorcarbons and chloninated hydrocarbons; slightly soluble in alcohol and insoluble in water. Milky yellow suspension at 200 mg plus 4 ml of water.


Bitte beachten Research Use Only (RUO).

Formel: C₃₆H₆₀O₂
Molecular Weight: 524,87 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00019414
CAS-Nummer: 79-81-2

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard
Activity ≥250000 USP u/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR