4-Aminoantipyrin, gelbes Pulver

Lieferant: MP Biomedicals
Achtung

Synonyme: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-on, 4-Aminophenazon

0215034310 0215034380 0215034383
ICNA0215034310EA 41.6 EUR
ICNA0215034310 ICNA0215034380 ICNA0215034383
4-Aminoantipyrin, gelbes Pulver
4-Aminoantipyrin

4-Aminoantipyrine is a reagent for determination of chlorinated phenols, phenylglycosidase, and glucose in the presence of phenol and peroxidase.


Soluble in water, ethanol, benzene; sparingly soluble in ether.


Bitte beachten Research Use Only (RUO).

Formel: C₁₁H₁₃N₃O
Molecular Weight: 203,24 g/mol
MDL: MFCD00003145
CAS-Nummer: 83-07-8

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard
Melting Point 104 - 114 °C
UV/VIS: λmax (water) Record Result

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR