o-Phenylendiamin
Lieferant: MP Biomedicals
Synonyme:
1,2-Diaminobenzol, 1,2-Phenylendiamin, OPD
0215182850
0215182883
ICNA0215182850EA
69.2
EUR
ICNA0215182850
ICNA0215182883
o-Phenylendiamin
o-Phenylendiamin
Formel:
C₆H₈N₂ Molecular Weight: 108,14 g/mol |
MDL:
MFCD00007721 CAS-Nummer: 95-54-5 UN: 1673 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...