Hydroxy naphthol blue disodium salt ACS

Lieferant: MP Biomedicals
Achtung

Synonyme: Hydroxynaphthol blue sodium salt, Hydroxy napthol blue, Sodium (E)-3-hydroxy-4-((2-hydroxy-6-sulpho-3-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-1-sulphonate

0215252910 0215252980
ICNA0215252910EA 34.6 EUR
ICNA0215252910 ICNA0215252980
Hydroxy naphthol blue disodium salt ACS
Hydroxy naphthol blue disodium salt

Hydroxynaphthol blue, disodium salt is used for determining the endpoint in metal complexation titrations.


Hydroxynaphthol blue, disodium salt is used for determining the endpoint in metal complexation titrations.


Soluble in water (clear, red solution).

Formel: C₂₀H₁₂N₂O₁₁S₃·2Na
Molecular Weight: 598,479 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004075
CAS-Nummer: 165660-27-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR