Brillantgrün (Malachitgrün G), grünes Pulver C.I. 42040
Lieferant: MP Biomedicals
Achtung
Synonyme: Ethyl Green, CI 42040, Basic Green 1, N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphate, Diamond Green, Malachite Green G, Emerald Green, Astradiamant green GX, Solid Green JO, Brilliant Green hydrogen sulphate
0215051625 0215051680
ICNA0215051625EA 31.9 EUR
ICNA0215051625 ICNA0215051680
Brillantgrün (Malachitgrün G), grünes Pulver C.I. 42040
Brillantgrün (Malachitgrün G)
Brilliant Green is one of the triarylmethane dyes. It is closely related to malachite green.
Soluble in water (1 mg/ml), ethanol, methanol.
Bitte beachten Research Use Only (RUO).
Formel: C₂₇H₃₄N₂O₄S
Molecular Weight: 482,65 g/mol
MDL: MFCD00011880
CAS-Nummer: 633-03-4
Bestellen
Spezifikation Testergebnisse
Identity Test Passes
UV/VIS: λmax (water) 625 ± 2 nm