Stärke, weißes Pulver ACS

Lieferant: MP Biomedicals

0215258280 0215258290 0215258291
ICNA0215258280EA 27.1 EUR
ICNA0215258280 ICNA0215258290 ICNA0215258291
Stärke, weißes Pulver ACS
Starch

Starch is used as an additive for food processing, food starches are typically used as thickeners and stabilisers.


Starch is used to produce various bioplastics, synthetic polymers that are biodegradable. An example is polylactic acid. Glucose from starch can be further fermented to biofuel corn ethanol. Hydrogen production can use starch as the raw material.


Soluble in water.


Bitte beachten Research Use Only (RUO).

Formel: (C₆H₁₀O₅)n
Molecular Weight: 162,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00082026
CAS-Nummer: 9005-25-8

Bestellen

Spezifikation Testergebnisse

Solubility Passes
pH (2% aq soln) 5,0 - 7,0
Residue on Ignition ≤0,4%
Sensitivity Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR