Guanosin-5'-triphosphat Trinatriumsalz, weißes Pulver

Lieferant: MP Biomedicals

Synonyme: (((2R,3S,4R,5R)-5-(2-Amino-6-oxo-1H-purin-9(6H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl)triphosphoric acid trisodium salt, (((2R,3S,4R,5R)-5-(2-Amino-6-oxo-1H-purin-9(6H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl)triphosphoric acid, trisodium salt, GTP trisodium salt

0215121610 0215121625 0215121680 0215121690
ICNA0215121610EA 36.2 EUR
ICNA0215121610 ICNA0215121625 ICNA0215121680 ICNA0215121690
Guanosin-5'-triphosphat Trinatriumsalz, weißes Pulver
Guanosin-5'-triphosphat Trinatriumsalz

Soluble in water (50 mg/ml - clear solution).


GTP functions as a carrier of phosphates and pyrophosphates involved in channeling chemical energy into specific biosynthetic pathways. GTP activates the signal transducing G proteins which are involved in various cellular processes including proliferation, differentiation, and activation of several intracellular kinase cascades. Proliferation and apoptosis are regulated in part by the hydrolysis of GTP by small GTPases Ras and Rho. Another type of small GTPase, Rab, plays a role in the docking and fusion of vesicles and may also be involved in vesicle formation. In addition to its role in signal transduction, GTP also serves as an energy-rich precursor of mononucleotide units in the enzymatic biosynthesis of DNA and RNA.


Soluble in water (50 mg/ml - clear solution).


Bitte beachten Research Use Only (RUO).

Formel: C₁₀H₁₃N₅Na₃O₁₄P₃
Molecular Weight: 588,94 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00077781
CAS-Nummer: 36051-31-7

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
UV/VIS: λmax (water) 250 - 255 nm
OD250/OD26 1.20 ± 0.0
OD280/OD260 0.65 ± 0.0
OD290/OD260 0.25 ± 0.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR