Tannin, braunes Pulver
Lieferant: MP Biomedicals
Synonyme: Gerbsäure, Tannin
0219027583 0219027590 0219027591
ICNA0219027583EA 58.3 EUR
ICNA0219027583 ICNA0219027590 ICNA0219027591
Tannin, braunes Pulver
Tannin
Tannic Acid is used for enzyme immobilization and protein adsorption
Tannic acid is used for enzyme immobilisation and protein adsorption.
Soluble in water (2 g/10 ml - clear solution), glycerol, alcohol, acetone. Practically insoluble in benzene, chloroform, ether, petroleum ether, carbondisulphide, carbon tetrachloride. Also soluble in ethanol (100 mg/ml - yellow to brown solution).
Bitte beachten Research Use Only (RUO).
Formel: C₇₆H₅₂O₄₆
Molecular Weight: 1701.2 g/mol
Flammpunkt: 199
Lagertemperatur: Raumtemperatur
MDL: MFCD00066397
CAS-Nummer: 1401-55-4
Merck Index: 12,09221
Bestellen
Spezifikation Testergebnisse
Identity Test Passes
Moisture ≤12%
FTIR Conforms to Standard