2-Mercaptoethansulfonsäure Natriumsalz (Mesna), weißes Pulver

Lieferant: MP Biomedicals
Achtung

Synonyme: Sodium 2-mercaptoethanesulphonate

0215160201 0215160205 0215160210 0215160225
ICNA0215160201EA 49 EUR
ICNA0215160201 ICNA0215160205 ICNA0215160210 ICNA0215160225
2-Mercaptoethansulfonsäure Natriumsalz (Mesna), weißes Pulver
2-Mercaptoethansulfonsäure Natriumsalz (Mesna)

2-Mercaptoethane sulphonic acid sodium salt is a thermally stable acidic cofactor in the methyl transfer reaction of methanobacterium.


Also acts as a water-soluble disulfide reducing agent.


Soluble in water (50 mg/ml).


Bitte beachten Research Use Only (RUO).

Formel: C₂H₅NaO₃S₂
Molecular Weight: 164,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007535
CAS-Nummer: 19767-45-4

Bestellen

Spezifikation Testergebnisse

Identity Test Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR