Trazodone hydrochloride, weißes Pulver

Lieferant: MP Biomedicals
Achtung

0219151801 0219151805
ICNA0219151801EA 56.9 EUR
ICNA0219151801 ICNA0219151805
Trazodone hydrochloride, weißes Pulver
Trazodone hydrochloride

Trazodone hydorchloride is a serotonin uptake inhibitor.


Antidepressant that potentiates the activity of serotonin uptake blockers and has full 5-HT2C serotonin receptor agonist activity. It is metabolized to the 5-HT1 serotonin receptor agonist 1-(3-Chlorophenyl)piperazine.


Soluble in chloroform; 0.1 M HCl(7.4 mg/ml) sparingly soluble in ethanol and methanol(25 mg/ml), DMSO; practically insoluble in common organic solvents. Insoluble in water.


Bitte beachten Research Use Only (RUO).

Formel: C₁₉H₂₃Cl₂N₅O
Schmelzpunkt: 223 °C (Lit.)
MDL: MFCD00079603
CAS-Nummer: 25332-39-2

Bestellen

Spezifikation Testergebnisse

Identity Test Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR