Weißes Mineralöl (Erdöl)

Lieferant: MP Biomedicals

ICNA0215169490EA 33 EUR
ICNA0215169490 ICNA0215169491
Weißes Mineralöl (Erdöl)
Mineral oil, white (petroleum)

Useful for preparation of nujol mulls for infrared spectroscopy.


Soluble in hydrocarbons; negligible soluble in water.

Lagertemperatur: Raumtemperatur
MDL: MFCD00131611
CAS-Nummer: 8042-47-5

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR