Polyvinylpyrrolidon, M.W. 40000 avg, weißes Pulver

Lieferant: MP Biomedicals

Synonyme: Polyvidon, Poly[1-(2-oxo-1-pyrrolidinyl)ethylen], PVP

0219545180 0219545190 0219545191
ICNA0219545180EA 55.8 EUR
ICNA0219545180 ICNA0219545190 ICNA0219545191
Polyvinylpyrrolidon, M.W. 40000 avg, weißes Pulver
Polyvinylpyrrolidon

Polyvinylpyrrolidone is a water-soluble polymer made from the monomer N-vinylpyrrolidone. Typically used as thickeners, dispersing agents, detoxicant, complexing agent, lubricants and binders.


Polyvinylpyrrolidone is used as an emulsifier and disintegrant for solution polymerisation; also for production of membranes, such as dialysis and water purification filters; as an aid for increasing the solubility of drugs in liquid and semi-liquid dosage forms (syrups, soft gelatine capsules) and as an inhibitor of recrystallisation.


Complexes with phenolics and alkaloids for their removal from plant samples, thus preventing their modification of proteins and any interference they may cause in spectrophotometric determinations of protein content. This is also reported to improve stability of enzymes.


Solubility ( 2,5% aq soln): clearly soluble, free of floating particles.


Bitte beachten Research Use Only (RUO).

Formel: (C₆H₉NO)n
Siedepunkt: 90…93 °C
Schmelzpunkt: 13,9 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01076626
CAS-Nummer: 9003-39-8

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard
pH (5% aq soln) 2.8 - 4.2
Solubility ( 2.5% aq soln) Clearly Soluble, Free of Floating Particles

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR