Spectinomycin dihydrochloride pentahydrate

Lieferant: MP Biomedicals

Synonyme: (2R,4aR,5aR,6S,7S,8R,9S,9aR,10aS)-4a,7,9-Trihydroxy-2-methyl-6,8-bis(methylamino)decahydro-4H-benzo[b]pyrano[2,3-e][1,4]dioxin-4-one dihydrochloride pentahydrate

0215206701 0215206725
ICNA0215206701EA 32.9 EUR
ICNA0215206701 ICNA0215206725
Spectinomycin dihydrochloride pentahydrate
Spectinomycin dihydrochloride pentahydrate

Soluble in water, methanol, propyleneglycol; very soluble benzene, chloroform, ethanol and acetone.

Formel: C₁₄H₂₄N₂O₇·2HCl·5H₂O
Molecular Weight: 495,35 g/mol
MDL: MFCD00150886
CAS-Nummer: 22189-32-8

Bestellen

Spezifikation Testergebnisse

Synonyms U18409AE; Actinospectacin; M141
Form White Powder
Storage Store at +4 °C
Heavy Metals ≤0,005%
Loss on Drying 20%
Optical Rotation Positive
Potency ≥500 μg/mg
Source Streptomyces sp.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR