Sudan III, rötlich braunes Pulver C.I. 26100
Lieferant: MP Biomedicals
Synonyme: Sudan Rot, Sudan Red BK, Sudan® III (Rot), 1-(4-(Phenylazo)phenylazo)-2-naphthol, Tony Red, Scarlet B, 1-([4-[(Phenyl)diazenyl]phenyl]diazenyl)naphthalen-2-ol, Fat Ponceau G, Sudan G, Cerasin Red, Sudan Red B, Organol® Rot BS, Sudan Red III, Sudan V
0215208725 0215208780
ICNA0215208725EA 107 EUR
ICNA0215208725 ICNA0215208780
Sudan III, rötlich braunes Pulver C.I. 26100
Sudan III
Sudan III is used in Coloring oils, sprit, lacquers etc; also as stain for zoological, pathological and vegetable objects like wax, cutin, resin, contents of lactiferous ducts etc, which are colored red, while cellulose membranes remain uncolored. Approved by FDA for external use only.
Insoluble in water; soluble in chloroform, glacial acetic acid; moderately soluble in alcohol, ether, acetone, fixed and volatile oils, hot glycerol. Clear red solution at 1 mg/ml in toluence.
Bitte beachten Research Use Only (RUO).
Formel: C₂₂H₁₆N₄O
Molecular Weight: 352,4 g/mol
Schmelzpunkt: 195 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003905
CAS-Nummer: 85-86-9
Bestellen
Spezifikation Testergebnisse
Identity Test Passes
UV/Vis: λmax (acetone) 502 - 510 nm