Tetramisole hydrochloride, Pulver

Lieferant: MP Biomedicals
Achtung

Synonyme: 6-Phenyl-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole hydrochloride, Tetramisole HCl, 6-Phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole hydrochloride

0215211905 0215211910
ICNA0215211905EA 54.6 EUR
ICNA0215211905 ICNA0215211910
Tetramisole hydrochloride, Pulver
Tetramisole hydrochloride

Tetramisole is an alkaline phosphatase inhibitor. It is a racemic mixture of (+) and (-) isomers. The (-) isomer (levamisole) accounts for most of the biological activity of tetramisole.


Suitable for inhibition of various mammalian alkaline phosphatases (i.e., liver, kidney, placenta, bone and tumor). Intestinal alkaline phosphatases are only slightly inhibited. Structure-activity relationship for tetramisole derivatives as alkaline phosphatase inhibitors. In addition to its use in enzyme and protein phosphorylation studies, tetramisole has been used to study membrane, tissue and animal systems.Tetramisole is an antihelminthic agent used in veterinary applications to treat helminth or worm infections.


Soluble in water (50 mg/ml - clear colorless solution).


Bitte beachten Research Use Only (RUO).

Formel: C₁₁H₁₃ClN₂S
Molecular Weight: 240.749 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005536
CAS-Nummer: 5086-74-8

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Melting Point 260 - 270 °C
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR