5-Aminosalicylsäure ≥99%, Pulver

Lieferant: MP Biomedicals

Synonyme: Mesalamine, Mesalazine, 5-Amino-2-hydroxybenzoic acid, m-Aminosalicylic acid

0215477025 0215477080
ICNA0215477025EA 153 EUR
ICNA0215477025 ICNA0215477080
5-Aminosalicylsäure ≥99%, Pulver
5-Aminosalicylsäure

5-Aminosalicylic acid is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is brown in color and can be read spectrophotometrically at 450 nm. The reaction may be stopped with 3 N NaOH and read at 550 nm .


Solubility: 200 mg plus 4 ml of 1 M HCl. Slightly soluble in cold water and alcohol. More soluble in hot water.

Formel: C₇H₇NO₃
Molecular Weight: 153,14 g/mol
MDL: MFCD00007877
CAS-Nummer: 89-57-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR