Weißer Ton (Kaolin), weißes Pulver
Lieferant: MP Biomedicals
Synonyme: Bolus Alba, Porzellanerde, Porzellanton, hydratisiertes Aluminiumsilikat, Pfeifenerde
0215514490 0215514491
ICNA0215514490EA 45.1 EUR
ICNA0215514490 ICNA0215514491
Weißer Ton (Kaolin), weißes Pulver
Weißer Ton (Kaolin)
Kaolin, also called china clay, soft white clay that is an essential ingredient in the manufacture of china and porcelain and is widely used in the making of paper, rubber, paint, and many other products.
Kaolins are used in a multiplicity of industries because of unique physical and chemical properties. Shape, particle size, color, softness, and non-abrasiveness are physical properties that are especially important. Chemical properties, such as comparatively low base exchange capacity, as well as ether chemical properties of the kaolin surface, and relative insolubility, are governing in many uses.
Formel: Al₂SiO₇·2H₂O
Molecular Weight: 258.16 g/mol
Schmelzpunkt: 1760
MDL: MFCD00062311
CAS-Nummer: 1332-58-7
Merck Index: 13,05300