Probenecid, weißes Pulver

Lieferant: MP Biomedicals
Achtung

Synonyme: 4-[(dipropylamino)sulphonyl]benzoic acid

0215637025 0215637080
ICNA0215637025EA 92.7 EUR
ICNA0215637025 ICNA0215637080
Probenecid, weißes Pulver
Probenecid

Probenecid is useful as an inhibitor of the organic anion transporter.


Probenecid is an inhibitor of multidrug resistance-associated proteins (Mdr-1). This compound blocks the active site of Mdr-1 and prevents cellular efflux of other active compounds, enhancing their effectiveness by allowing them to remain in the cytoplasm.


Soluble in dilute alkalies; slightly soluble ether and acetone; sparingly soluble in chloroform and ethanol; practically insoluble in water and dilute acids.

Formel: C₁₃H₁₉NO₄S
Molecular Weight: 285,4 g/mol
Schmelzpunkt: 381…385 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00038402
CAS-Nummer: 57-66-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR