5-(N-2-3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis(2,3-dihydroxypropyl)isophthalamide, weißes Pulver
Lieferant: MP Biomedicals
Synonyme:
Iohexol, N1,N3-Bis(2,3-dihydroxypropyl)-5-(N-(2,3-dihydroxypropyl)acetamido)-2,4,6-triiodoisophthalamide, 5-[Acetyl(2,3-dihydroxypropyl)amino]-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide, N,N'-Bis(2,3-dihydroxypropyl)-5-[N-(2,3-dihydroxypropyl)acetamido]-2,4,6-triiodoisophthalamide
Non-ionic monomeric triiodinated x-ray contrast agent
5-(N-2,3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis-(2,3-dihydroxypropyl)isophthalamide is useful as a nonionic density gradient medium.
Density gradient medium to measure hemodynamic parameters in vivo. Induces apoptosis. Orally active.
Soluble in water (50 mg/ml). It is also soluble in formamide and dimethyformamide.
Formel:
C₁₉H₂₆I₃N₃O₉ Molecular Weight: 821,142 g/mol |
MDL:
MFCD00077732 CAS-Nummer: 66108-95-0 |
Spezifikation Testergebnisse
Identity Test | Passes |
FTIR | Conforms to Reference |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...