Ammoniumeisen(III)citrat, braunes Pulver

Lieferant: MP Biomedicals
Achtung

Synonyme: Ammonium ferric citrate, Ferric Ammonium Citrate, Ammonium iron (III) citrate

0215804080 0215804090
ICNA0215804080EA 24.6 EUR
ICNA0215804080 ICNA0215804090
Ammoniumeisen(III)citrat, braunes Pulver
Ammoniumeisen(III)citrat

Ferric Ammonium Citrate is supplemented to the growth media as a source of iron for the micro-organism to be analysed.


  • Soluble in water (100 mg/ml); practically insoluble in alcohol.

Formel: C₆H₅+4y)Fe(x)N(y)O₇
Siedepunkt: 2735 °C (1013 hPa) (Lit.)
Schmelzpunkt: 1539 °C (Lit.)
Dichte: 7.87 g/cm³ (25 °C) (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013099
CAS-Nummer: 1185-57-5

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard
Iron Content 16.5 - 18.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR