Zincon sodium salt, braunes Pulver
Lieferant: MP Biomedicals
|
Achtung
|
Zincon is a colorimetric indicator for Zn, Cu and Hg. Reagent for the spectrophotometric determination of zinc. Simultaneous determination of zinc and copper. Used in automated flow injection analysis of zinc. As a metal indicator for EDTA titration of Zn, or for the back titration of Ca in the presence of Mg with glycoletherdiaminetertraacetic acid (GEDTA or EGTA).
Reacts with Zn (pH 8,5 to 9,5) and Cu (pH 5,0 to 9,5) to form blue chelates. Also color reaction occurs with Co, Ni and some other polyvalent cations, Beer's law is observed in the range of 0,1 to 2,4 μg Cu/ml in water at 600 mu.
Slightly soluble in water and alcohol. Insoluble in most organic solvents. Soluble in aqueous alkali resulting in a red solution.
Formel:
C₂₀H₁₅N₄NaO₆S Schmelzpunkt: 250…260 °C (Zers., Lit.) |
MDL:
MFCD00064385 CAS-Nummer: 62625-22-3 |
Spezifikation Testergebnisse
Identity Test | Passes |
Storage | Store at Room Temperature (15 - 30 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...