2-Methyl-4-isothiazolin-3-one hydrochloride ≥99%, klare bis weißgraue Lösung
Lieferant: MP Biomedicals
Gefahr
Synonyme: MIT
0215952290 0215952291
ICNA0215952290EA 41.9 EUR
ICNA0215952290 ICNA0215952291
2-Methyl-4-isothiazolin-3-one hydrochloride ≥99%, klare bis weißgraue Lösung
2-Methyl-4-isothiazolin-3-one hydrochloride
2-Methylisothiazolone (MIT) are chemical compounds used as highly effective biocidal agents against fungal and bacterial species. They are widely used as a preservative in cosmetic industry, and as a pesticide, e.g. in paints and water tanks.
  • UV/Visible Absorbance: A405: 0.002(lit.)
  • pH: pH optimum is between pH 5 and 7.5
  • Pb: 1 ppm(lit.)
2-Methylisothiazolone (MIT) shows a broad efficiency spectrum against gram-positive and gram-negative bacteria as well as yeast and fungi. It can be used alone or in combination with bromo-nitro-dioxane. In contrasts to sodium azide MIT is not mutagenic, is considerably less toxic and performs outstanding biocidic properties at low application concentrations.
MIT is used to study the effects of tyrosine phosphorylation on focal adhesion kinase (FAK) activity in the development of neural axons and dendrites.
Soluble in water, methanol and ethanol (96%).
Formel: C₄H₆ClNOS
Molecular Weight: 151.608 g/mol
Schmelzpunkt: ~ 175 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD06804636
CAS-Nummer: 26172-54-3
UN: 1759
ADR: 8 II
Bestellen
Spezifikation Testergebnisse
Identity Test Passes
Purity ≥99%
FTIR Conforms to Reference