Chlorhexidindi(acetat) ≥97%, weißes Pulver
Lieferant: MP Biomedicals
|
Gefahr
|
Synonyme:
1,1'-Hexamethylenebis[5-(p-chlorophenyl)biguanide] diacetate, 1,1'-Hexamethylenebis[5-(4-chlorophenyl)biguanide] Diacetate
An antiseptic and disinfectant agent. It is effective against a wide range of bacteria, some fungi and some viruses, and an agent for the prevention of gingivitis. Commercial ophthalmic products have used this agent to replace thimerosal as a preservative; however, it can cause skin irritation.
- Critical Micelle Concentration (CMC): 0,01% (w/v, 25 °C)(Lit.)
Chlorhexidine is used primarily as a topical antiseptic/disinfectant in wound healing, at catheterization sites, in various dental applications and in surgical scrubs. It has been used to study the inactivation of HIV and the immobilization of sperm. It was used to study effective skin antisepsis and disinfection of medical devices as well as the effects of antimicrobials on dental plaque.
Chlorhexidine is a cationic broad-spectrum antimicrobial agent belonging to the bis(biguanide) family. Its mechanism of action involves destabilization of the outer bacterial membrane. It is effective against both Gram-positive and Gram-negative bacteria. Chlorhexidine is used primarily as a topical antiseptic/disinfectant in wound healing, at catheterization sites, in various dental applications and in surgical scrubs.
Soluble in water (0,15 g/10 ml water with heat) , in ethanol (50 mg/ml - clear, faint yellow solution), alcohol, glycerol, propylene, glycol, polyethylene glycols.
A solution of 0,5% in water has been used for general disinfection. Used in conjuction with cetyltrimethylammonium bromide (CTAB) can increase its effectiveness.
Formel:
C₂₂H₃₀Cl₂N₁₀ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012532 CAS-Nummer: 56-95-1 UN: 3077 ADR: 9,III |
Spezifikation Testergebnisse
Identity Test | Passes |
Purity | ≥97% |
FTIR | Conforms to Standard |
Melting Point | 151 - 159 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...