Brilliant sulphaflavine, dunkeloranges Pulver

Lieferant: MP Biomedicals

0219507805 0219507825
ICNA0219507805EA 55.2 EUR
ICNA0219507805 ICNA0219507825
Brilliant sulphaflavine, dunkeloranges Pulver
Brilliant sulphaflavine

Brilliant Sulfaflavine is not the same as FIAT (Brilliant Sulfoflavin FF; Acid Yellow 7) which has only one methyl group para to the nitrogen.


  • Concentration: Dye Content: ∼ 40% (Lit.)
  • Refractive Index: ~1.75 (Predicted) (lit.)


Brilliant sulfaflavine was used as fluorescent tracers to study the application rate of foliar fungicides when applied to soybeans for the control of rust.


Soluble in water; very slightly soluble in ethanol.

Formel: C₁₉H₁₃N₂NaO₅S
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2391-30-2

Bestellen

Spezifikation Testergebnisse

Identity Test Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR