Bromkresolpurpur, Pulver
Lieferant: MP Biomedicals
Achtung
Synonyme: 5′,5''-Dibrom-o-kresolsulfonphthalein
0219508005 0219508025 0219508080
ICNA0219508005EA 45.5 EUR
ICNA0219508005 ICNA0219508025 ICNA0219508080
Bromkresolpurpur, Pulver
Bromkresolpurpur
Bromocresol Purple is used as a pH indicator (5.2-6.8 : yellow to blue-purple). Used in spectrophotometric determination of serum-albumin.
Bromocresol purple binds quantitatively with human serum albumin forming a stable complex with absorbance maximum at 600 nm. The intensity of the color produced is directly proportional to the albumin concentration in the sample.
Soluble in water (20 mg/ml), ethanol (80 mg/ml) or methyl cellosolve (300 mg/ml). Soluble in methanol (4 mg/ml).
Formel: C₂₁H₁₆Br₂O₅S
Molecular Weight: 540,23 g/mol
Schmelzpunkt: 240 °C (Zers., Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011681
CAS-Nummer: 115-40-2
Bestellen
Spezifikation Testergebnisse
Identity Test Passes
UV/Vis: l max (MeOH) 418 ± 3 nm
pH Color Interval Passes