Cobalt(II)chlorid Hexahydrat, rötlich violette Kristalle
Lieferant: MP Biomedicals
Gefahr
Synonyme: Cobaltdichlorid Hexahydrat
ICNA0219510783EA 114 EUR
ICNA0219510783 ICNA0219510780 ICNA0219510725
Cobalt(II)chlorid Hexahydrat, rötlich violette Kristalle
Cobalt(II)chlorid Hexahydrat
Cobalt chloride is utilized in commercial applications such as electroplating, catalyst preparation, painting on glass and porcelain, and in the manufacture of vitamin B12.
  • Key Applications : Indicator for water in dessicants
  • Application Areas : Chemistry
  • Product Type : Biochemicals
  • Biochemical Category : Labels, Dyes & Stains
  • Chemical Class : Labels, Dyes & Stains
  • Vapor Pressure : 40 mm Hg(Lit.)
  • Presentation : Redish Purple Crystals
  • Format : Crystals
  • Solubility : Soluble in water (100 mg/mL), alcohols, acetone, ether, glycerol.
It is also used as a humidity and water indicator; when cobalt chloride is used with desiccants, it will change from blue to pink upon absorption of moisture. Cobalt chloride is also used as an additive in fertilizer and feed.
Solubility : Soluble in water (100 mg/ml), alcohols, acetone, ether, glycerol.
Formel: CoCl₂·6H₂O
Molecular Weight: 237,93 g/mol
Dichte: 1,920 g/mL (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149652
CAS-Nummer: 7791-13-1
UN: 3077
ADR: 6.1,III
Bestellen
Spezifikation Testergebnisse
Identity Test Passes
pH (0.2 M aq soln) Report Result