n-Hexadecan ≥99%, Flüssigkeit
Lieferant: MP Biomedicals
Synonyme: Cetan
0219521880 0219521890 0219521891
ICNA0219521880EA 66.4 EUR
ICNA0219521880 ICNA0219521890 ICNA0219521891
n-Hexadecan ≥99%, Flüssigkeit
n-Hexadecan
Hexadecane has been used as a phase change material (PCM) in the preparation of hexadecane/xGnP (exfoliated graphite nanoplatelet), a composite PCM loaded with xGnP.
Soluble in ether; insoluble in water.
Transition temp.- solidification point 17.5 - 18.5 °C (Lit.)
Formel: H₃C(CH₂)₁₄CH₃
Molecular Weight: 226.45 g/mol
Siedepunkt: 287
Schmelzpunkt: 18
Flammpunkt: 135
MDL: MFCD00008998
CAS-Nummer: 544-76-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90d3084bdd908b305ecf9b3c30e03cd98a407d94&vid=e5a15f4f6c36024a34521e08083957252ff78457&prid=web&clid=SAPDAM