D(+)-Saccharose, Kristalle, für die Nahrungsmittelindustrie
Lieferant: MP Biomedicals
Synonyme:
Haushaltszucker, D(+)-Sucrose, Rübenzucker, Rohrzucker, 2-β-D-Fructofuranosyl-1-α-D-glucopyranosid, Sucrose, Kristallzucker
0290471305
0290471325
0290471380
ICNA0290471305EA
49.5
EUR
ICNA0290471305
ICNA0290471325
ICNA0290471380
D(+)-Saccharose, Kristalle, für die Nahrungsmittelindustrie
D(+)-Saccharose
Soluble in water (1 g/0,5 ml at 25 °C; 1 g/0,2 ml boiling water), ethanol (1 g/170 ml) or methanol (1 g/100 ml); moderately soluble in glycerol or pyridine.
Formel:
C₁₂H₂₂O₁₁ Molecular Weight: 342.3 g/mol Schmelzpunkt: 177…187 °C |
MDL:
MFCD00006626 CAS-Nummer: 57-50-1 Merck Index: 13,08966 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...