Gummi Xanthan Xanthomonas campestris, powder_off-white
Lieferant: MP Biomedicals
Synonyme: Xanthan Gummi, Xanthan
0296002180 0296002190
ICNA0296002180EA 53.7 EUR
ICNA0296002180 ICNA0296002190
Gummi Xanthan Xanthomonas campestris, powder_off-white
Xanthan gum
Xanthan gum is a diet suspending agent that will maintain insolubles in a homogenous suspended state for at least 24 hours.
Xanthan gum is a useful matrix component for drug delivery systems. It forms stable drug suspensions in aqueous media and soft gels with locust bean gum or guar gum.
Readily dissolves in water with stirring to give highly vicous solutions at low concentrations.
MDL: MFCD00131256
CAS-Nummer: 11138-66-2
Bestellen
Spezifikation Testergebnisse
Identity Test Passes
Loss on Drying Record Result