Anti-GITRL Mouse Monoclonal Antibody

Lieferant: Novus Biologicals
NOVUNBP2-11965EA 614 EUR
NOVUNBP2-11965
Anti-GITRL Mouse Monoclonal Antibody
Antikörper
Mouse Monoclonal GITRL Antibody (MM0312-4Q25). Tested Applications: Blocking/Neutralizing, Flow Cytometry. Tested Reactivity: Human.

Type: Primary
Antigen: GITRL
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR