Anti-LAG3 Mouse Monoclonal Antibody

Lieferant: Novus Biologicals
NOVUNBP1-97662EA 510 EUR
NOVUNBP1-97662
Anti-LAG3 Mouse Monoclonal Antibody
Antikörper
Mouse Monoclonal Lymphocyte Activation Gene 3 Antibody (11E3). Tested Applications: Western Blot, Blocking/Neutralizing, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Frozen, Immunoprecipitation. Tested Reactivi

Type: Primary
Antigen: LAG3
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR