Anti-BCL2A1 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NOVUNBP1-76715EA 689 EUR
NOVUNBP1-76715
Anti-BCL2A1 Rabbit Polyclonal Antibody
Antikörper
Rabbit Polyclonal Bcl2A1 Antibody. Tested Applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse.

Type: Primary
Antigen: BCL2A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR