Inlet kits for Agilent GC, FastPack™
Lieferant: Restek
21101 21102 21104
REST21101EA 320 EUR
REST21101 REST21102 REST21104
Inlet kits for Agilent GC, FastPack™
Chromatographie-Zubehör Chromatographie-Ersatzteile
These FastPack™ inlet kits for Agilent GC make routine injection port maintenance easy.
  • Convenient - all the parts provided in one package
  • Economical - costs less than the sum of the individual parts
  • Clean - Mylar bag is factory sealed and no contamination of the products from weeks in the lab
Lieferumfang: Each kit is supplied with one each, inlet liner, Viton® O-ring, 0,8 mm ID gold-plated inlet seal, inlet seal washer and 11 mm Thermolite® septum.