Nortriptylin -hydrochlorid, weißes Pulver
Lieferant: MP Biomedicals
|
Achtung
|
Synonyme:
3-(10,11-Dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N-methyl-1-propanamine Hydrochloride, 10,11-Dihydro-5-(N-methyl-3-aminopropylidene)-5H-dibenzo[a,d]cycloheptene Hydrochloride
Nortriptyline Hydrochloride is a second-generation tricyclic antidepressant.
Nortriptyline hydrochloride is a more potent inhibitor of the norepinephrine transporter (Ki = 3.4 nM) than the serotonin transporter (Ki = 161 nM). 5-HT2 serotonin receptor antagonist.
Soluble in water, chloroform; sparingly soluble in methal, ethanol (100 mg/mL); practically insoluble in ether, benzene.
Bitte beachten Research Use Only (RUO).
Formel:
C₁₉H₂₁N Molecular Weight: 299,84 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00058024 CAS-Nummer: 894-71-3 |
Spezifikation Testergebnisse
Identity Test | Passes |
Melting Point | 213 - 217 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...