Trifluoressigsäureanhydrid
Lieferant: Restek
Gefahr
|
Synonyme:
Trifluoracetic anhydride, TFAA
Acylation derivatization reagent.
- Most commonly used for electron capture detection
- React with alcohols, amines, and phenols
- Frequently used for drugs of abuse confirmation
Acylation reagents offer the same advantage available from silylation reagents: creating less polar, more volatile derivatives. In comparison to silylating reagents, the acylating reagents can more readily target highly polar multifunctional compounds, such as carbohydrates and amino acids. In addition, acylating reagents offer the distinct advantage of introducing electron-capturing groups, thus enhancing detectability during analysis.
Formel:
C₄F₆O₃ Molecular Weight: 210,03 g/mol |
MDL:
MFCD00000416 CAS-Nummer: 407-25-0 UN: 3265 ADR: 8,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...